Assay Details
Gene Symbol: Mat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145569.4 | NP_663544.1 | 2 - 3 | 285 | 72 | |
GenBank mRNA | AB070266.1 | - | 2 - 3 | 249 | 72 | |
AK034757.1 | - | 2 - 3 | 286 | 72 | ||
AK052764.1 | - | 2 - 3 | 284 | 72 | ||
AK079420.1 | - | 2 - 3 | 291 | 72 | ||
AK133960.1 | - | 2 - 3 | 284 | 72 | ||
AK144189.1 | - | 2 - 3 | 288 | 72 | ||
AK145407.1 | - | 2 - 3 | 284 | 72 | ||
AK150040.1 | - | 2 - 3 | 284 | 72 | ||
AK151281.1 | - | 2 - 3 | 284 | 72 | ||
AK160869.1 | - | 2 - 3 | 284 | 72 | ||
AK161852.1 | - | 2 - 3 | 284 | 72 | ||
AK164137.1 | - | 2 - 3 | 284 | 72 | ||
AK166671.1 | - | 2 - 3 | 284 | 72 | ||
AK168155.1 | - | 2 - 3 | 284 | 72 | ||
AK169705.1 | - | 2 - 3 | 284 | 72 | ||
AK190931.1 | - | 2 - 3 | 114 | 72 | ||
AK206552.1 | - | 2 - 3 | 114 | 72 | ||
AK206774.1 | - | 2 - 3 | 114 | 72 | ||
AK210321.1 | - | 1 - 2 | 61 | 72 | ||
AK213948.1 | - | 2 - 3 | 114 | 72 | ||
AK216312.1 | - | 2 - 3 | 114 | 72 | ||
BC003451.1 | - | 2 - 3 | 225 | 2655444 | 72 | |
BC058360.1 | - | 2 - 3 | 266 | 6821781 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
