Assay Details
Gene Symbol: Mat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145569.4 | NP_663544.1 | 3 - 4 | 409 | 78 | |
GenBank mRNA | AK034757.1 | - | 3 - 4 | 410 | 78 | |
AK052764.1 | - | 3 - 4 | 408 | 78 | ||
AK079420.1 | - | 3 - 4 | 415 | 78 | ||
AK133960.1 | - | 3 - 4 | 408 | 78 | ||
AK144189.1 | - | 3 - 4 | 412 | 78 | ||
AK145407.1 | - | 3 - 4 | 408 | 78 | ||
AK150040.1 | - | 3 - 4 | 408 | 78 | ||
AK160869.1 | - | 3 - 4 | 408 | 78 | ||
AK161852.1 | - | 3 - 4 | 408 | 78 | ||
AK164137.1 | - | 3 - 4 | 408 | 78 | ||
AK166671.1 | - | 3 - 4 | 408 | 78 | ||
AK168155.1 | - | 3 - 4 | 408 | 78 | ||
AK190931.1 | - | 3 - 4 | 238 | 78 | ||
AK200511.1 | - | 3 - 4 | 315 | 78 | ||
AK206552.1 | - | 3 - 4 | 238 | 78 | ||
AK206774.1 | - | 3 - 4 | 238 | 78 | ||
AK210321.1 | - | 2 - 3 | 185 | 78 | ||
AK213948.1 | - | 3 - 4 | 238 | 78 | ||
AK215611.1 | - | 3 - 4 | 238 | 78 | ||
AK216312.1 | - | 3 - 4 | 238 | 78 | ||
AK219100.1 | - | 3 - 4 | 238 | 78 | ||
BC003451.1 | - | 3 - 4 | 349 | 2655444 | 78 | |
BC058360.1 | - | 3 - 4 | 390 | 6821781 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
