Assay Details
Gene Symbol: Aak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040106.2 | NP_001035195.1 | 7 - 8 | 1404 | 103 | |
NM_177762.6 | NP_808430.2 | 7 - 8 | 1404 | 103 | ||
GenBank mRNA | AK147363.1 | - | 7 - 8 | 1405 | 103 | |
AK158879.1 | - | 7 - 8 | 1294 | 103 | ||
BC141176.1 | - | 7 - 8 | 1160 | 9055788 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
