Assay Details
Gene Symbol: Copg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017477.2 | NP_059505.1 | 7 - 8 | 589 | 78 | |
GenBank mRNA | AF187079.1 | - | 7 - 8 | 588 | 78 | |
AK049417.1 | - | 7 - 8 | 540 | 78 | ||
AK077461.1 | - | 7 - 8 | 536 | 78 | ||
AK147895.1 | - | 7 - 8 | 602 | 78 | ||
AK151284.1 | - | 7 - 8 | 594 | 78 | ||
AK151816.1 | - | 7 - 8 | 594 | 78 | ||
AK153142.1 | - | 7 - 8 | 594 | 78 | ||
AK164280.1 | - | 7 - 8 | 559 | 78 | ||
AK169256.1 | - | 7 - 8 | 634 | 78 | ||
AK171950.1 | - | 7 - 8 | 568 | 78 | ||
AK172519.1 | - | 7 - 8 | 605 | 78 | ||
BC024686.1 | - | 7 - 8 | 560 | 3992144 | 78 | |
BC024896.1 | - | 7 - 8 | 557 | 4189311 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
