Assay Details
Gene Symbol: Rasgef1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027526.1 | NP_081802.1 | 3 - 4 | 409 | 69 | |
GenBank mRNA | AK018120.1 | - | 2 - 3 | 240 | 69 | |
BC138284.1 | - | 3 - 4 | 375 | 8861303 | 69 | |
BC145232.1 | - | 3 - 4 | 375 | 9053784 | 69 | |
BC145752.1 | - | 3 - 4 | 375 | 40131055 | 69 | |
BY256830.1 | - | 3 - 4 | 357 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
