Assay Details
Gene Symbol: Rasgef1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027526.1 | NP_081802.1 | 4 - 5 | 544 | 73 | |
GenBank mRNA | AK018120.1 | - | 3 - 4 | 375 | 73 | |
AK158756.1 | - | 5 - 6 | 570 | 73 | ||
BC138284.1 | - | 4 - 5 | 510 | 8861303 | 73 | |
BC145752.1 | - | 4 - 5 | 510 | 40131055 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
