Assay Details
Gene Symbol: Rasgef1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027526.1 | NP_081802.1 | 5 - 6 | 763 | 52 | |
GenBank mRNA | AK018120.1 | - | 4 - 5 | 594 | 52 | |
AK158756.1 | - | 6 - 7 | 789 | 52 | ||
BC138284.1 | - | 5 - 6 | 729 | 8861303 | 52 | |
BC145232.1 | - | 5 - 6 | 756 | 9053784 | 52 | |
BC145752.1 | - | 5 - 6 | 729 | 40131055 | 52 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
