Assay Details
Gene Symbol: Rasgef1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027526.1 | NP_081802.1 | 9 - 10 | 1117 | 75 | |
GenBank mRNA | AK018120.1 | - | 8 - 9 | 948 | 75 | |
AK158756.1 | - | 10 - 11 | 1143 | 75 | ||
BC138284.1 | - | 9 - 10 | 1083 | 8861303 | 75 | |
BC145232.1 | - | 9 - 10 | 1110 | 9053784 | 75 | |
BC145752.1 | - | 9 - 10 | 1083 | 40131055 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
