Assay Details
Gene Symbol: Rasgef1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027526.1 | NP_081802.1 | 10 - 11 | 1302 | 71 | |
GenBank mRNA | AK018120.1 | - | 9 - 10 | 1133 | 71 | |
AK158756.1 | - | 11 - 12 | 1328 | 71 | ||
BC138284.1 | - | 10 - 11 | 1268 | 8861303 | 71 | |
BC145232.1 | - | 10 - 11 | 1295 | 9053784 | 71 | |
BC145752.1 | - | 10 - 11 | 1268 | 40131055 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
