Assay Details
Gene Symbol: Rtkn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009106.2 | NP_033132.2 | 1 - 2 | 372 | 64 | |
GenBank mRNA | CX218519.1 | - | 1 - 2 | 372 | 64 | |
U54638.1 | - | 1 - 2 | 375 | 64 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_009106.2, U54638.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
