Assay Details
Gene Symbol: Rtkn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136227.1 | NP_001129699.1 | 5 - 6 | 689 | 71 | |
NM_009106.2 | NP_033132.2 | 6 - 7 | 1096 | 71 | ||
NM_133641.2 | NP_598396.2 | 5 - 6 | 1600 | 71 | ||
GenBank mRNA | AK076155.1 | - | 5 - 6 | 610 | 71 | |
BC004824.1 | - | 5 - 6 | 930 | 71 | ||
BC013820.1 | - | 5 - 6 | 1574 | 3981927 | 71 | |
U54638.1 | - | 6 - 7 | 1099 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
