Assay Details
Gene Symbol: Actg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009610.2 | NP_033740.2 | 1 - 2 | 75 | 78 | |
GenBank mRNA | AK146631.1 | - | 1 - 2 | 48 | 78 | |
AK168649.1 | - | 1 - 2 | 48 | 78 | ||
BC002042.1 | - | 1 - 2 | 56 | 3591323 | 78 | |
BY778654.1 | - | 1 - 2 | 76 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK146631.1, AK168649.1, BC002042.1, NM_009610.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
