Assay Details
Gene Symbol: Actg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009610.2 | NP_033740.2 | 6 - 7 | 718 | 98 | |
GenBank mRNA | AK146631.1 | - | 6 - 7 | 691 | 98 | |
AK168649.1 | - | 6 - 7 | 691 | 98 | ||
BC002042.1 | - | 6 - 7 | 699 | 3591323 | 98 | |
M26689.1 | - | 5 - 6 | 647 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
