Assay Details
Gene Symbol: P3h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013534.4 | NP_038562.2 | 1 - 2 | 495 | 65 | |
GenBank mRNA | AJ441086.1 | - | 1 - 2 | 495 | 65 | |
AY463530.1 | - | 1 - 2 | 495 | 65 | ||
BC003726.1 | - | 1 - 2 | 81 | 3497610 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC003726.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
