Assay Details
Gene Symbol: P3h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013534.4 | NP_038562.2 | 2 - 3 | 651 | 63 | |
GenBank mRNA | AJ441086.1 | - | 2 - 3 | 651 | 63 | |
AK140962.1 | - | 2 - 3 | 694 | 63 | ||
AY463530.1 | - | 2 - 3 | 651 | 63 | ||
BC003726.1 | - | 2 - 3 | 237 | 3497610 | 63 | |
BC016431.1 | - | 1 - 2 | 137 | 3483280 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
