Assay Details
Gene Symbol: Usp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013700.1 | NP_038728.1 | 14 - 15 | 1814 | 81 | |
GenBank mRNA | AK154008.1 | - | 14 - 15 | 1803 | 81 | |
AK179736.1 | - | 2 - 3 | 235 | 81 | ||
AK194987.1 | - | 2 - 3 | 235 | 81 | ||
BC066993.1 | - | 14 - 15 | 1792 | 5689229 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
