Assay Details
Gene Symbol: Clstn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153508.4 | NP_705728.1 | 6 - 7 | 1230 | 69 | |
GenBank mRNA | AJ278486.1 | - | 6 - 7 | 1097 | 69 | |
AK032336.1 | - | 6 - 7 | 1172 | 69 | ||
BC041781.1 | - | 6 - 7 | 929 | 69 | ||
BC055054.1 | - | 6 - 7 | 1162 | 6493096 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
