Assay Details
Gene Symbol: Aim1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162970.1 | NP_001156442.1 | 11 - 12 | 3591 | 59 | |
GenBank mRNA | AK145690.1 | - | 11 - 12 | 3597 | 59 | |
BC042709.1 | - | 6 - 7 | 612 | 59 | ||
BC150738.1 | - | 10 - 11 | 3520 | 9087649 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
