Assay Details
Gene Symbol: Cnksr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081047.1 | NP_001074516.1 | 8 - 9 | 841 | 97 | |
GenBank mRNA | AK145910.1 | - | 8 - 9 | 803 | 97 | |
BC113805.1 | - | 8 - 9 | 806 | 40062208 | 97 | |
BC113956.1 | - | 8 - 9 | 806 | 40062209 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
