Assay Details
Gene Symbol: Aim1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162970.1 | NP_001156442.1 | 18 - 19 | 4411 | 74 | |
GenBank mRNA | AK145690.1 | - | 18 - 19 | 4418 | 74 | |
AK165037.1 | - | 4 - 5 | 2372 | 74 | ||
BC042709.1 | - | 13 - 14 | 74 | |||
BC077737.1 | - | 1 - 2 | 418 | 74 | ||
BC150738.1 | - | 17 - 18 | 4340 | 9087649 | 74 | |
BP759371.1 | - | 2 - 3 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
