Assay Details
Gene Symbol: Eya4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010167.4 | NP_034297.2 | 11 - 12 | 1247 | 101 | |
GenBank mRNA | AK041625.1 | - | 11 - 12 | 1248 | 101 | |
BC120898.1 | - | 11 - 12 | 1231 | 40061597 | 101 | |
BC120899.1 | - | 11 - 12 | 1231 | 40061599 | 101 | |
Y17115.1 | - | 12 - 13 | 1358 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
