Assay Details
Gene Symbol: Aim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172393.2 | NP_765981.2 | 10 - 11 | 4619 | 125 | |
GenBank mRNA | AF397913.1 | - | 4 - 5 | 326 | 125 | |
AK041082.1 | - | 9 - 10 | 1235 | 125 | ||
AK088102.1 | - | 9 - 10 | 1291 | 125 | ||
BC035551.1 | - | 9 - 10 | 1771 | 125 | ||
BC059272.1 | - | 9 - 10 | 1776 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
