Assay Details
Gene Symbol: Kpna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008468.4 | NP_032494.3 | 2 - 3 | 215 | 94 | |
GenBank mRNA | AF020773.1 | - | 1 - 2 | 119 | 94 | |
AK046371.1 | - | 2 - 3 | 174 | 94 | ||
AK084212.1 | - | 2 - 3 | 173 | 94 | ||
AK154882.1 | - | 2 - 3 | 200 | 94 | ||
BC004833.1 | - | 1 - 2 | 131 | 2651272 | 94 | |
CJ082540.1 | - | 2 - 3 | 236 | 94 | ||
CT010260.1 | - | 1 - 2 | 119 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
