Assay Details
Gene Symbol: Kpna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008468.4 | NP_032494.3 | 8 - 9 | 839 | 66 | |
GenBank mRNA | AF020773.1 | - | 7 - 8 | 743 | 66 | |
AK046371.1 | - | 8 - 9 | 798 | 66 | ||
AK084212.1 | - | 8 - 9 | 797 | 66 | ||
AK154882.1 | - | 8 - 9 | 824 | 66 | ||
BC004833.1 | - | 7 - 8 | 755 | 2651272 | 66 | |
CT010260.1 | - | 7 - 8 | 743 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
