Assay Details
Gene Symbol: Ece1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK197559.1 | - | 2 - 3 | 183 | 96 | |
AK202528.1 | - | 2 - 3 | 96 | |||
BC038057.1 | - | 6 - 7 | 666 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
