Assay Details
Gene Symbol: Thoc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172438.3 | NP_766026.1 | 12 - 13 | 1228 | 62 | |
GenBank mRNA | AK006840.1 | - | 3 - 4 | 236 | 62 | |
AK050734.1 | - | 12 - 13 | 1228 | 62 | ||
AK173078.1 | - | 10 - 11 | 1066 | 62 | ||
BC020332.1 | - | 11 - 12 | 1193 | 62 | ||
BC021779.1 | - | 11 - 12 | 1202 | 62 | ||
BC039758.1 | - | 12 - 13 | 1229 | 3995737 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
