Assay Details
Gene Symbol: Stk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009288.2 | NP_033314.2 | 3 - 4 | 505 | 84 | |
GenBank mRNA | AK148118.1 | - | 3 - 4 | 448 | 84 | |
AK220448.1 | - | 3 - 4 | 427 | 84 | ||
BC069840.1 | - | 3 - 4 | 450 | 84 | ||
BC096624.1 | - | 3 - 4 | 462 | 84 | ||
BC128363.1 | - | 3 - 4 | 391 | 40111177 | 84 | |
D89728.1 | - | 3 - 4 | 466 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
