Assay Details
Gene Symbol: Stk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009288.2 | NP_033314.2 | 4 - 5 | 656 | 73 | |
GenBank mRNA | AK148118.1 | - | 4 - 5 | 599 | 73 | |
AK220448.1 | - | 4 - 5 | 578 | 73 | ||
BC069840.1 | - | 4 - 5 | 601 | 73 | ||
BC096624.1 | - | 4 - 5 | 613 | 73 | ||
BC128363.1 | - | 4 - 5 | 542 | 40111177 | 73 | |
D89728.1 | - | 4 - 5 | 617 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
