Assay Details
Gene Symbol: Tnfaip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166402.1 | NP_001159874.1 | 4 - 5 | 992 | 65 | |
NM_009397.3 | NP_033423.3 | 5 - 6 | 1077 | 65 | ||
GenBank mRNA | AK088297.1 | - | 5 - 6 | 1034 | 65 | |
AK151921.1 | - | 5 - 6 | 1038 | 65 | ||
AK153238.1 | - | 5 - 6 | 1032 | 65 | ||
AK172529.1 | - | 4 - 5 | 994 | 65 | ||
BC054806.1 | - | 5 - 6 | 1077 | 6838303 | 65 | |
BC060221.1 | - | 5 - 6 | 1077 | 6843338 | 65 | |
CT010397.1 | - | 4 - 5 | 804 | 65 | ||
U19463.1 | - | 4 - 5 | 804 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
