Assay Details
Gene Symbol: Tnfaip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166402.1 | NP_001159874.1 | 5 - 6 | 1177 | 95 | |
NM_009397.3 | NP_033423.3 | 6 - 7 | 1262 | 95 | ||
GenBank mRNA | AK088297.1 | - | 6 - 7 | 1219 | 95 | |
AK151921.1 | - | 6 - 7 | 1223 | 95 | ||
AK153238.1 | - | 6 - 7 | 1217 | 95 | ||
AK171326.1 | - | 5 - 6 | 1176 | 95 | ||
AK172529.1 | - | 5 - 6 | 1179 | 95 | ||
BC050259.1 | - | 5 - 6 | 2359 | 95 | ||
BC054806.1 | - | 6 - 7 | 1262 | 6838303 | 95 | |
BC060221.1 | - | 6 - 7 | 1262 | 6843338 | 95 | |
CT010397.1 | - | 5 - 6 | 989 | 95 | ||
U19463.1 | - | 5 - 6 | 989 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
