Assay Details
Gene Symbol: Pdzrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018884.2 | NP_061372.2 | 1 - 2 | 733 | 107 | |
GenBank mRNA | AF127084.1 | - | 1 - 2 | 733 | 107 | |
AF127085.1 | - | 2 - 3 | 577 | 107 | ||
AK135776.1 | - | 1 - 2 | 178 | 107 | ||
AK143226.1 | - | 1 - 2 | 247 | 107 | ||
AK173098.1 | - | 1 - 2 | 692 | 107 | ||
HM560981.1 | - | 1 - 2 | 724 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
