Assay Details
Gene Symbol: Pex5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277805.1 | NP_001264734.1 | 1 - 2 | 95 | 73 | |
NM_175933.2 | NP_787947.1 | 1 - 2 | 95 | 73 | ||
GenBank mRNA | AJ416473.1 | - | 1 - 2 | 77 | 73 | |
AK161470.1 | - | 1 - 2 | 92 | 73 | ||
BC029748.1 | - | 1 - 2 | 65 | 2599159 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AJ416473.1, AK161470.1, BC029748.1, NM_001277805.1, NM_175933.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
