Assay Details
Gene Symbol: Il17re | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034029.1 | NP_001029201.1 | 12 - 13 | 1207 | 82 | |
NM_001034031.1 | NP_001029203.1 | 14 - 15 | 1282 | 82 | ||
NM_145826.5 | NP_665825.2 | 13 - 14 | 1634 | 82 | ||
GenBank mRNA | AK046302.1 | - | 13 - 14 | 1637 | 82 | |
AK046713.1 | - | 13 - 14 | 1296 | 82 | ||
AK078876.1 | - | 5 - 6 | 1548 | 82 | ||
AK169498.1 | - | 13 - 14 | 1462 | 82 | ||
BC026737.1 | - | 3 - 4 | 378 | 82 | ||
DQ092339.1 | - | 13 - 14 | 1223 | 82 | ||
DQ092340.1 | - | 12 - 13 | 1207 | 82 | ||
DQ092341.1 | - | 14 - 15 | 1282 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
