Assay Details
Gene Symbol: Grid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008167.2 | NP_032193.1 | 3 - 4 | 528 | 96 | |
GenBank mRNA | AK036192.1 | - | 3 - 4 | 559 | 96 | |
AK141070.1 | - | 1380 | 96 | |||
BC139823.1 | - | 3 - 4 | 532 | 40141838 | 96 | |
D13266.1 | - | 3 - 4 | 528 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
