Assay Details
Gene Symbol: Cd27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033126.2 | NP_001028298.1 | 3 - 4 | 582 | 66 | |
GenBank mRNA | AK153895.1 | - | 3 - 4 | 521 | 66 | |
AK154005.1 | - | 3 - 4 | 566 | 66 | ||
BC139125.1 | - | 3 - 4 | 547 | 8862147 | 66 | |
BC139126.1 | - | 3 - 4 | 547 | 8862148 | 66 | |
BE631582.1 | - | 2 - 3 | 221 | 66 | ||
L24495.1 | - | 3 - 4 | 624 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
