Assay Details
Gene Symbol: Hace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172473.3 | NP_766061.2 | 2 - 3 | 377 | 93 | |
GenBank mRNA | AK042879.1 | - | 2 - 3 | 318 | 93 | |
AK156958.1 | - | 2 - 3 | 319 | 93 | ||
BC120695.1 | - | 2 - 3 | 274 | 40129618 | 93 | |
BC120697.1 | - | 2 - 3 | 274 | 40129620 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
