Assay Details
Gene Symbol: 2310069B03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040520.1 | 1 - 2 | 84 | 155 | ||
GenBank mRNA | AK078808.1 | - | 1 - 2 | 85 | 155 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | hexokinase 2 | Hk2 |
AI642394, HKII |
-106.8kb | Sense | Chr. 6 - 82725025 - 82774454 on NCBI Build GRCm38 |
Downstream |
sema domain, immunoglobulin domain (Ig), TM domain, and short cytoplasmic domain |
Sema4f | Sema W | 30.0kb | Sense | Chr. 6 - 82911884 - 82939761 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
