Assay Details
Gene Symbol: Atp2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177236.4 | NP_796210.2 | 11 - 12 | 2195 | 68 | |
GenBank mRNA | AK032322.1 | - | 11 - 12 | 2120 | 68 | |
AK045516.1 | - | 11 - 12 | 2195 | 68 | ||
BC118975.1 | - | 10 - 11 | 1927 | 40098391 | 68 | |
BC118976.1 | - | 10 - 11 | 1927 | 40098398 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
