Assay Details
Gene Symbol: Esr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AI386103.1 | - | 2 - 3 | 265 | 88 | |
AK039911.1 | - | 4 - 5 | 618 | 88 | ||
AK041525.1 | - | 2 - 3 | 263 | 88 | ||
AK139745.1 | - | 2 - 3 | 675 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK041525.1, AK139745.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
