Assay Details
Gene Symbol: Abcg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | KF662865.1 | - | 1 - 2 | 43 | 69 | |
KF662866.1 | - | 1 - 2 | 43 | 69 | ||
KF662867.1 | - | 1 - 2 | 43 | 69 | ||
KF662869.1 | - | 1 - 2 | 43 | 69 | ||
KF662870.1 | - | 1 - 2 | 43 | 69 | ||
KF662871.1 | - | 1 - 2 | 43 | 69 |
Gene Symbol: 4930430M16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK015257.1 | - | 1 - 2 | 46 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the KF662865.1,KF662866.1,KF662867.1,KF662869.1,KF662870.1,KF662871.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
