Assay Details
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025392.1 | NP_001020563.1 | 8 - 9 | 2284 | 71 | |
NM_001025393.1 | NP_001020564.1 | 8 - 9 | 2284 | 71 | ||
NM_153787.2 | NP_722482.1 | 8 - 9 | 2278 | 71 | ||
GenBank mRNA | AK088741.1 | - | 8 - 9 | 2219 | 71 | |
AK129071.1 | - | 5 - 6 | 1568 | 71 | ||
AK141834.1 | - | 8 - 9 | 2231 | 71 | ||
AK149724.1 | - | 8 - 9 | 2237 | 71 | ||
AK150017.1 | - | 8 - 9 | 2237 | 71 | ||
AK161608.1 | - | 1173 | 71 | |||
AK161692.1 | - | 8 - 9 | 2282 | 71 | ||
BC029682.1 | - | 8 - 9 | 2215 | 71 | ||
BC034300.1 | - | 8 - 9 | 2210 | 4507876 | 71 | |
BC086624.1 | - | 8 - 9 | 2266 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
