Assay Details
Gene Symbol: Limk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010718.4 | NP_034848.1 | 2 - 3 | 296 | 82 | |
GenBank mRNA | AB005131.1 | - | 2 - 3 | 149 | 82 | |
AB008117.1 | - | 2 - 3 | 232 | 82 | ||
BC007129.1 | - | 2 - 3 | 239 | 3496836 | 82 | |
BY253265.1 | - | 2 - 3 | 296 | 82 | ||
U88618.1 | - | 2 - 3 | 149 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
