Assay Details
Gene Symbol: Tspan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133681.4 | NP_598442.1 | 5 - 6 | 582 | 82 | |
GenBank mRNA | AK007428.1 | - | 5 - 6 | 580 | 82 | |
AK078869.1 | - | 5 - 6 | 583 | 82 | ||
AK162521.1 | - | 5 - 6 | 582 | 82 | ||
BC003448.1 | - | 5 - 6 | 522 | 2654254 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
