Assay Details
Gene Symbol: Ust | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177387.3 | NP_796361.2 | 1 - 2 | 672 | 101 | |
GenBank mRNA | AK086181.1 | - | 1 - 2 | 619 | 101 | |
AK147850.1 | - | 1 - 2 | 673 | 101 | ||
AK162950.1 | - | 1 - 2 | 245 | 101 | ||
BC138155.1 | - | 1 - 2 | 412 | 8861174 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
