Assay Details
Gene Symbol: Ust | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177387.3 | NP_796361.2 | 5 - 6 | 1114 | 125 | |
GenBank mRNA | AK086181.1 | - | 5 - 6 | 1061 | 125 | |
AK147850.1 | - | 5 - 6 | 1115 | 125 | ||
AK162950.1 | - | 5 - 6 | 687 | 125 | ||
BC138155.1 | - | 5 - 6 | 854 | 8861174 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
