Assay Details
Gene Symbol: Npc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207242.2 | NP_997125.2 | 10 - 11 | 2762 | 69 | |
GenBank mRNA | AK078947.1 | - | 10 - 11 | 2663 | 69 | |
AY437866.1 | - | 10 - 11 | 2640 | 69 | ||
BC131788.1 | - | 10 - 11 | 2639 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
