Assay Details
Gene Symbol: Npc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207242.2 | NP_997125.2 | 1 - 2 | 173 | 58 | |
GenBank mRNA | AK078947.1 | - | 1 - 2 | 72 | 58 | |
AY437866.1 | - | 1 - 2 | 51 | 58 | ||
BC131788.1 | - | 1 - 2 | 50 | 58 | ||
BC131789.1 | - | 1 - 2 | 51 | 40086103 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
