Assay Details
Gene Symbol: Ddc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190448.1 | NP_001177377.1 | 12 - 13 | 1285 | 146 | |
NM_016672.4 | NP_057881.1 | 12 - 13 | 1223 | 146 | ||
GenBank mRNA | AF071068.1 | - | 12 - 13 | 1156 | 146 | |
AK149141.1 | - | 12 - 13 | 1217 | 146 | ||
BC060989.1 | - | 12 - 13 | 1241 | 146 | ||
BC089594.1 | - | 13 - 14 | 1226 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
