Assay Details
Gene Symbol: Ros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011282.2 | NP_035412.2 | 23 - 24 | 3724 | 94 | |
GenBank mRNA | U15443.1 | - | 23 - 24 | 3723 | 94 | |
X81650.1 | - | 23 - 24 | 3547 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
