Assay Details
Gene Symbol: Stx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016797.4 | NP_058077.2 | 2 - 3 | 559 | 79 | |
GenBank mRNA | AF056323.1 | - | 2 - 3 | 171 | 79 | |
AK002867.1 | - | 2 - 3 | 228 | 79 | ||
AK013869.1 | - | 2 - 3 | 213 | 79 | ||
AK082927.1 | - | 2 - 3 | 199 | 79 | ||
AK142742.1 | - | 206 | 79 | |||
AK158923.1 | - | 2 - 3 | 560 | 79 | ||
AK166822.1 | - | 2 - 3 | 229 | 79 | ||
AK169822.1 | - | 2 - 3 | 232 | 79 | ||
AK210437.1 | - | 1 - 2 | 71 | 79 | ||
BC132123.1 | - | 1 - 2 | 109 | 40130400 | 79 | |
BC132125.1 | - | 1 - 2 | 109 | 40130402 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
